(1R,6S)-8-Cbz-3,8-diazabicyclo[4.2.0]octane
Supplier: Apollo Scientific
OR470481-1G OR470481-250MG
APOSOR470481-1GEA 2860 EUR
APOSOR470481-1G APOSOR470481-250MG
(1R,6S)-8-Cbz-3,8-diazabicyclo[4.2.0]octane
(1R,6S)-8-Cbz-3,8-diazabicyclo[4.2.0]octane
Formula: C₁₄H₁₈N₂O₂
MW: 246.305 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22205852
CAS Number: 1932802-76-0