3-(3-Pyridyl)-2-propyn-1-ol 98%
Supplier: Apollo Scientific
Warning
Synonyms: 3-(Pyridin-3-yl)prop-2-yn-1-ol
OR470565-5G OR470565-10G OR470565-1G
APOSOR470565-5GEA 393 EUR
APOSOR470565-5G APOSOR470565-10G APOSOR470565-1G
3-(3-Pyridyl)-2-propyn-1-ol 98%
3-(3-Pyridyl)-2-propyn-1-ol
Formula: C₈H₇NO
MW: 133.147 g/mol
Melting Pt: 97…100 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00168869
CAS Number: 61266-33-9