Ethyl Imidazo[1,2-a]pyridine-5-carboxylate
Supplier: Apollo Scientific
Synonyms: Ethyl imidazo[1,2-a]pyridine-5-carboxylate
OR470583-1G OR470583-5G OR470583-250MG
APOSOR470583-1GEA 448 EUR
APOSOR470583-1G APOSOR470583-5G APOSOR470583-250MG
Ethyl Imidazo[1,2-a]pyridine-5-carboxylate
Ethyl Imidazo[1,2-a]pyridine-5-carboxylate
Formula: C₁₀H₁₀N₂O₂
MW: 190.199 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13176744
CAS Number: 177485-39-1