3-(2-Pyridyl)-2-propyn-1-ol 98%
Supplier: Apollo Scientific
Warning
Synonyms: 3-(Pyridin-2-yl)prop-2-yn-1-ol
OR470619-5G OR470619-10G OR470619-1G OR470619-25G
APOSOR470619-5GEA 271 EUR
APOSOR470619-5G APOSOR470619-10G APOSOR470619-1G APOSOR470619-25G
3-(2-Pyridyl)-2-propyn-1-ol 98%
3-(2-Pyridyl)-2-propyn-1-ol
Formula: C₈H₇NO
MW: 133.147 g/mol
Melting Pt: 83…85 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00168870
CAS Number: 29768-03-4