2-Bromo-4,6-dimethoxybenzaldehyde
Supplier: Apollo Scientific
OR470723-5G OR470723-1G
APOSOR470723-5GEA 511 EUR
APOSOR470723-5G APOSOR470723-1G
2-Bromo-4,6-dimethoxybenzaldehyde
2-Bromo-4,6-dimethoxybenzaldehyde
Formula: C₉H₉BrO₃
Storage Temperature: Ambient
MDL Number: MFCD11110636
CAS Number: 81574-69-8