2-(Methoxymethyl)phenol

Supplier: Apollo Scientific

OR470757-5G OR470757-1G
APOSOR470757-5GEA 325 EUR
APOSOR470757-5G APOSOR470757-1G
2-(Methoxymethyl)phenol
2-(Methoxymethyl)phenol
Formula: C₈H₁₀O₂
MW: 138.164 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14680991
CAS Number: 5635-98-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR