5,7-Dimethylpyrazolo[1,5-a]pyrimidine-3-carboxylic acid
Supplier: Apollo Scientific
OR470775-5G OR470775-1G
APOSOR470775-5GEA 184 EUR
APOSOR470775-5G APOSOR470775-1G
5,7-Dimethylpyrazolo[1,5-a]pyrimidine-3-carboxylic acid
5,7-Dimethylpyrazolo[1,5-a]pyrimidine-3-carboxylic acid
Formula: C₉H₉N₃O₂
MW: 191.187 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01051014
CAS Number: 90349-23-8