N-Isobutylcyclopropanamine

Supplier: Apollo Scientific

OR470797-1G OR470797-5G
APOSOR470797-1GEA 397 EUR
APOSOR470797-1G APOSOR470797-5G
N-Isobutylcyclopropanamine
N-Isobutylcyclopropanamine
Formula: C₇H₁₅N
MW: 113.201 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09047821
CAS Number: 215523-02-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR