Methyl 3,4-Dihydro-2H-pyran-5-carboxylate
Supplier: Apollo Scientific
Warning
OR470823-5G OR470823-1G
APOSOR470823-5GEA 804 EUR
APOSOR470823-5G APOSOR470823-1G
Methyl 3,4-Dihydro-2H-pyran-5-carboxylate
Methyl 3,4-Dihydro-2H-pyran-5-carboxylate
Formula: C₇H₁₀O₃
MW: 142.152 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06203714
CAS Number: 86971-83-7