3'-(Chloromethyl)-2,6-dimethyl-4-[3-(methylsulphonyl)propoxy]biphenyl
Supplier: Apollo Scientific
OR470893-1G OR470893-250MG OR470893-5G
APOSOR470893-1GEA 1220 EUR
APOSOR470893-1G APOSOR470893-250MG APOSOR470893-5G
3'-(Chloromethyl)-2,6-dimethyl-4-[3-(methylsulphonyl)propoxy]biphenyl
3'-(Chloromethyl)-2,6-dimethyl-4-[3-(methylsulphonyl)propoxy]biphenyl
Formula: C₁₉H₂₃ClO₃S
MW: 366.902 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27959537
CAS Number: 1394138-49-8