Methyl 7-methoxy-4-oxo-1,4-dihydroquinoline-6-carboxylate
Supplier: Apollo Scientific
Warning
OR470912-5G
APOSOR470912-5GEA 78.3 EUR
APOSOR470912-5G
Methyl 7-methoxy-4-oxo-1,4-dihydroquinoline-6-carboxylate
Methyl 7-methoxy-4-oxo-1,4-dihydroquinoline-6-carboxylate
Formula: C₁₂H₁₁NO₄
MW: 233.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13192255
CAS Number: 205448-65-3