Methyl 3'-Hydroxybiphenyl-4-carboxylate
Supplier: Apollo Scientific
OR471006-1G OR471006-5G
APOSOR471006-1GEA 306 EUR
APOSOR471006-1G APOSOR471006-5G
Methyl 3'-Hydroxybiphenyl-4-carboxylate
Methyl 3'-Hydroxybiphenyl-4-carboxylate
Formula: C₁₄H₁₂O₃
MW: 228.243 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06409247
CAS Number: 579511-01-6