6-Chloro-5-cyanonicotinic acid
Supplier: Apollo Scientific
Warning
Synonyms: 6-Chloro-5-cyanonicotinic Acid
OR471009-1G OR471009-250MG OR471009-5G
APOSOR471009-1GEA 713 EUR
APOSOR471009-1G APOSOR471009-250MG APOSOR471009-5G
6-Chloro-5-cyanonicotinic acid
6-Chloro-5-cyanonicotinic acid
Formula: C₇H₃ClN₂O₂
MW: 182.564 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14585457
CAS Number: 1206969-20-1