4-Bromo-7-methylindole
Supplier: Apollo Scientific
Synonyms: 4-Bromo-7-methyl-1H-indole
OR471099-1G OR471099-250MG
APOSOR471099-1GEA 133 EUR
APOSOR471099-1G APOSOR471099-250MG
4-Bromo-7-methylindole
4-Bromo-7-methylindole
Formula: C₉H₈BrN
MW: 210.071 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08272256
CAS Number: 936092-87-4