3,6-Dimethyl-2,4-dinitrophenol
Supplier: Apollo Scientific
OR471100-100G OR471100-25G OR471100-5G
APOSOR471100-100GEA 1540 EUR
APOSOR471100-100G APOSOR471100-25G APOSOR471100-5G
3,6-Dimethyl-2,4-dinitrophenol
3,6-Dimethyl-2,4-dinitrophenol
Formula: C₈H₈N₂O₅
MW: 212.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01741407
CAS Number: 15968-56-6