4-(Cbz-amino)benzaldehyde
Supplier: Apollo Scientific
OR471203-1G OR471203-250MG OR471203-5G
APOSOR471203-1GEA 306 EUR
APOSOR471203-1G APOSOR471203-250MG APOSOR471203-5G
4-(Cbz-amino)benzaldehyde
4-(Cbz-amino)benzaldehyde
Formula: C₁₅H₁₃NO₃
Storage Temperature: Ambient
MDL Number: MFCD17170461
CAS Number: 71150-68-0