5-Bromo-2-furanmethanamine

Supplier: Apollo Scientific

OR471307-1G OR471307-250MG
APOSOR471307-1GEA 1540 EUR
APOSOR471307-1G APOSOR471307-250MG
5-Bromo-2-furanmethanamine
5-Bromo-2-furanmethanamine
Formula: C₅H₆BrNO
MW: 176.011 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08848187
CAS Number: 263169-37-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR