Quinoline-4-carbohydrazide Hydrate
Supplier: Apollo Scientific
OR471337-1G
OR471337-250MG
APOSOR471337-1GEA
1010
EUR
APOSOR471337-1G
APOSOR471337-250MG
Quinoline-4-carbohydrazide Hydrate
Quinoline-4-carbohydrazide Hydrate
Formula:
C₁₀H₉N₃O·H₂O MW: 205.213 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28968740 CAS Number: 2006277-54-7 |
Learn more
![](/m-si.vwr.com/sl_SI/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...