1-Boc-4-oxo-1-azaspiro[5.5]undecane
Supplier: Apollo Scientific
Warning
OR471347-1G OR471347-250MG OR471347-5G
APOSOR471347-1GEA 601 EUR
APOSOR471347-1G APOSOR471347-250MG APOSOR471347-5G
1-Boc-4-oxo-1-azaspiro[5.5]undecane
1-Boc-4-oxo-1-azaspiro[5.5]undecane
Formula: C₁₅H₂₅NO₃
MW: 267.364 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09283648
CAS Number: 778647-35-1