1,3-Dimethyl-5-hydroxyuracil
Supplier: Apollo Scientific
OR471490-1G OR471490-250MG OR471490-5G
APOSOR471490-1GEA 448 EUR
APOSOR471490-1G APOSOR471490-250MG APOSOR471490-5G
1,3-Dimethyl-5-hydroxyuracil
1,3-Dimethyl-5-hydroxyuracil
Formula: C₆H₈N₂O₃
MW: 156.139 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06658203
CAS Number: 408335-42-2