2-Hydroxy-7-methyl-1,4-naphthoquinone
Supplier: Apollo Scientific
OR471495-1G OR471495-250MG OR471495-5G
APOSOR471495-1GEA 601 EUR
APOSOR471495-1G APOSOR471495-250MG APOSOR471495-5G
2-Hydroxy-7-methyl-1,4-naphthoquinone
2-Hydroxy-7-methyl-1,4-naphthoquinone
Formula: C₁₁H₈O₃
MW: 188.179 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27925890
CAS Number: 58472-26-7