4'-Amino-2'-chloroacetophenone
Supplier: Apollo Scientific
OR471500-1G OR471500-250MG OR471500-5G
APOSOR471500-1GEA 367 EUR
APOSOR471500-1G APOSOR471500-250MG APOSOR471500-5G
4'-Amino-2'-chloroacetophenone
4'-Amino-2'-chloroacetophenone
Formula: C₈H₈ClNO
MW: 169.608 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11848240
CAS Number: 72531-23-8