4,6-Dibromonicotinaldehyde
Supplier: Apollo Scientific
OR471502-1G OR471502-250MG OR471502-5G
APOSOR471502-1GEA 367 EUR
APOSOR471502-1G APOSOR471502-250MG APOSOR471502-5G
4,6-Dibromonicotinaldehyde
4,6-Dibromonicotinaldehyde
Formula: C₆H₃Br₂NO
MW: 264.902 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18261845
CAS Number: 1211585-10-2