2-(Boc-amino)-4-methoxy-1-nitrobenzene
Supplier: Apollo Scientific
Synonyms: tert-Butyl (5-methoxy-2-nitrophenyl)carbamate
OR471530-1G OR471530-250MG
APOSOR471530-1GEA 252 EUR
APOSOR471530-1G APOSOR471530-250MG
2-(Boc-amino)-4-methoxy-1-nitrobenzene
2-(Boc-amino)-4-methoxy-1-nitrobenzene
Formula: C₁₂H₁₆N₂O₅
MW: 268.266 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28043790
CAS Number: 185428-55-1