O-(3-Buten-2-yl)hydroxylamine hydrochloride
Supplier: Apollo Scientific
Synonyms: O-(But-3-en-2-yl)hydroxylamine hydrochloride
OR471531-1G OR471531-250MG
APOSOR471531-1GEA 563 EUR
APOSOR471531-1G APOSOR471531-250MG
O-(3-Buten-2-yl)hydroxylamine hydrochloride
O-(3-Buten-2-yl)hydroxylamine hydrochloride
Formula: C₄H₉NO·ClH
MW: 123.581 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09838140
CAS Number: 71350-16-8