2-Aminopyrazolo[1,5-a]pyridine
Supplier: Apollo Scientific
OR471538-1G OR471538-250MG OR471538-5G
APOSOR471538-1GEA 713 EUR
APOSOR471538-1G APOSOR471538-250MG APOSOR471538-5G
2-Aminopyrazolo[1,5-a]pyridine
2-Aminopyrazolo[1,5-a]pyridine
Formula: C₇H₇N₃
MW: 133.151 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19208536
CAS Number: 51119-05-2