2-Amino-4-isopropyl-5-methylthiazole
Supplier: Apollo Scientific
OR471541-1G OR471541-5G OR471541-250MG
APOSOR471541-1GEA 397 EUR
APOSOR471541-1G APOSOR471541-5G APOSOR471541-250MG
2-Amino-4-isopropyl-5-methylthiazole
2-Amino-4-isopropyl-5-methylthiazole
Formula: C₇H₁₂N₂S
MW: 156.249 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12784032
CAS Number: 101012-43-5