4-(4-Pyridyl)-3-butyn-1-ol
Supplier: Apollo Scientific
Synonyms: 4-(Pyridin-4-yl)but-3-yn-1-ol
OR471582-1G OR471582-5G
APOSOR471582-1GEA 111 EUR
APOSOR471582-1G APOSOR471582-5G
4-(4-Pyridyl)-3-butyn-1-ol
4-(4-Pyridyl)-3-butyn-1-ol
Formula: C₉H₉NO
MW: 147.174 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18384538
CAS Number: 192643-83-7