5-Methoxy-4-[(4-methoxybenzyl)oxy]-2-nitrobenzoic acid 97%
Supplier: Apollo Scientific
Synonyms: 5-Methoxy-4-((4-methoxybenzyl)oxy)-2-nitrobenzoic acid
OR471691-10G OR471691-1G OR471691-5G
APOSOR471691-10GEA 112 EUR
APOSOR471691-10G APOSOR471691-1G APOSOR471691-5G
5-Methoxy-4-[(4-methoxybenzyl)oxy]-2-nitrobenzoic acid 97%
5-Methoxy-4-[(4-methoxybenzyl)oxy]-2-nitrobenzoic acid
Formula: C₁₆H₁₅NO₇
MW: 333.293 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28144964
CAS Number: 1643979-88-7