Methyl 4-chloro-2-methoxy-5-methylbenzoate 95%
Supplier: Apollo Scientific
OR471719-1G OR471719-250MG
APOSOR471719-1GEA 1200 EUR
APOSOR471719-1G APOSOR471719-250MG
Methyl 4-chloro-2-methoxy-5-methylbenzoate 95%
Methyl 4-chloro-2-methoxy-5-methylbenzoate
Formula: C₁₀H₁₁ClO₃
MW: 214.646 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31695764
CAS Number: 1368228-61-8