2-Isopentyl-1,3-dioxoisoindoline-5-carbaldehyde 95%
Supplier: Apollo Scientific
OR471720-1G OR471720-250MG
APOSOR471720-1GEA 1200 EUR
APOSOR471720-1G APOSOR471720-250MG
2-Isopentyl-1,3-dioxoisoindoline-5-carbaldehyde 95%
2-Isopentyl-1,3-dioxoisoindoline-5-carbaldehyde
Formula: C₁₄H₁₅NO₃
MW: 245.274 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31695765
CAS Number: 2243974-49-2