N-[1-(3-Bromophenyl)ethyl]cyclopropanamine hydrochloride
Supplier: Apollo Scientific
Warning
OR475076-1G OR475076-250MG
APOSOR475076-1GEA 133 EUR
APOSOR475076-1G APOSOR475076-250MG
N-[1-(3-Bromophenyl)ethyl]cyclopropanamine hydrochloride
N-[1-(3-Bromophenyl)ethyl]cyclopropanamine hydrochloride
Formula: C₁₁H₁₄BrN·ClH
MW: 276.601 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD31695691
CAS Number: 2367002-65-9