N-[1-(3-Bromophenyl)ethyl]prop-2-yn-1-amine hydrochloride
Supplier: Apollo Scientific
Warning
OR475107-1G OR475107-250MG
APOSOR475107-1GEA 126 EUR
APOSOR475107-1G APOSOR475107-250MG
N-[1-(3-Bromophenyl)ethyl]prop-2-yn-1-amine hydrochloride
N-[1-(3-Bromophenyl)ethyl]prop-2-yn-1-amine hydrochloride
Formula: C₁₁H₁₂BrN·ClH
MW: 274.585 g/mol
Storage Temperature: Ambient
MDL Number: MFCD31695640
CAS Number: 2367002-63-7