N-[1-(4-Bromophenyl)ethyl]prop-2-yn-1-amine hydrochloride
Supplier: Apollo Scientific
Warning
OR475108-1G OR475108-250MG
APOSOR475108-1GEA 126 EUR
APOSOR475108-1G APOSOR475108-250MG
N-[1-(4-Bromophenyl)ethyl]prop-2-yn-1-amine hydrochloride
N-[1-(4-Bromophenyl)ethyl]prop-2-yn-1-amine hydrochloride
Formula: C₁₁H₁₂BrN·ClH
MW: 274.585 g/mol
Storage Temperature: Freezer
MDL Number: MFCD31695643
CAS Number: 2387695-90-9