Methyl 4-bromo-1-cyclopropyl-1H-pyrazole-3-carboxylate
Supplier: Apollo Scientific
Warning
OR475185-1G OR475185-250MG
APOSOR475185-1GEA 313 EUR
APOSOR475185-1G APOSOR475185-250MG
Methyl 4-bromo-1-cyclopropyl-1H-pyrazole-3-carboxylate
Methyl 4-bromo-1-cyclopropyl-1H-pyrazole-3-carboxylate
Formula: C₈H₉BrN₂O₂
MW: 245.073 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32200418
CAS Number: 2407349-25-9