Methyl 4-chloro-1-cyclopropyl-1H-pyrazole-3-carboxylate
Supplier: Apollo Scientific
Warning
OR475186-1G OR475186-250MG
APOSOR475186-1GEA 313 EUR
APOSOR475186-1G APOSOR475186-250MG
Methyl 4-chloro-1-cyclopropyl-1H-pyrazole-3-carboxylate
Methyl 4-chloro-1-cyclopropyl-1H-pyrazole-3-carboxylate
Formula: C₈H₉ClN₂O₂
MW: 200.622 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32200419
CAS Number: 2407349-24-8