(1-Cyclopropyl-1H-pyrazol-3-yl)methanol
Supplier: Apollo Scientific
Warning
OR475187-1G OR475187-250MG
APOSOR475187-1GEA 522 EUR
APOSOR475187-1G APOSOR475187-250MG
(1-Cyclopropyl-1H-pyrazol-3-yl)methanol
(1-Cyclopropyl-1H-pyrazol-3-yl)methanol
Formula: C₇H₁₀N₂O
MW: 138.167 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32200420
CAS Number: 2201840-10-8