Methyl 3-bromo-5-chloropyridine-2-carboxylate
Supplier: Apollo Scientific
Warning
Synonyms: Methyl 3-bromo-5-chloropicolinate, 3-Bromo-5-chloro-2-(methoxycarbonyl)pyridine
OR47837-1G OR47837-250MG
APOSOR47837-1GEA 183 EUR
APOSOR47837-1G APOSOR47837-250MG
Methyl 3-bromo-5-chloropyridine-2-carboxylate
Methyl 3-bromo-5-chloropyridine-2-carboxylate
Formula: C₇H₅BrClNO₂
MW: 250.477 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14698105
CAS Number: 1214362-45-4