2-Methyl-5,6,7,8-tetrahydro-[1,2,4]triazolo[1,5-a]pyridin-6-amine
Supplier: Apollo Scientific
OR47920-100MG OR47920-1G OR47920-250MG
APOSOR47920-100MGEA 302 EUR
APOSOR47920-100MG APOSOR47920-1G APOSOR47920-250MG
2-Methyl-5,6,7,8-tetrahydro-[1,2,4]triazolo[1,5-a]pyridin-6-amine
2-Methyl-5,6,7,8-tetrahydro-[1,2,4]triazolo[1,5-a]pyridin-6-amine
Formula: C₇H₁₂N₄
MW: 152.197 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16817413
CAS Number: 1251925-31-1