3,4-Dichlorobenzenesulphonamide 97%
Supplier: Apollo Scientific
OR480672-100G OR480672-25G
APOSOR480672-100GEA 870 EUR
APOSOR480672-100G APOSOR480672-25G
3,4-Dichlorobenzenesulphonamide 97%
3,4-Dichlorobenzenesulphonamide
Formula: C₆H₅Cl₂NO₂S
MW: 226.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01314052
CAS Number: 23815-28-3