Quinoxaline-2,3-diamine 95%
Supplier: Apollo Scientific
OR480759-1G OR480759-5G
APOSOR480759-1GEA 127 EUR
APOSOR480759-1G APOSOR480759-5G
Quinoxaline-2,3-diamine 95%
Quinoxaline-2,3-diamine
Formula: C₈H₈N₄
MW: 160.176 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00193133
CAS Number: 6640-47-7