N-Methyl-7-nitro-1,2,3,4-tetrahydroquinoline 97%
Supplier: Apollo Scientific
Synonyms: 1-Methyl-7-nitro-1,2,3,4-tetrahydroquinoline
OR480784-1G OR480784-5G
APOSOR480784-1GEA 40.6 EUR
APOSOR480784-1G APOSOR480784-5G
N-Methyl-7-nitro-1,2,3,4-tetrahydroquinoline 97%
N-Methyl-7-nitro-1,2,3,4-tetrahydroquinoline
Formula: C₁₀H₁₂N₂O₂
MW: 192.214 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00496655
CAS Number: 39275-18-8