N-(4-Chlorophenyl)-1,2-phenylenediamine 99%
Supplier: Apollo Scientific
Warning
Synonyms: N1-(4-Chlorophenyl)benzene-1,2-diamine, 2-Amino-4′-chlorodiphenylamine, 2-(4-Chloroanilino)aniline
OR48122-25G OR48122-5G OR48122-100G
APOSOR48122-25GEA 58 EUR
APOSOR48122-25G APOSOR48122-5G APOSOR48122-100G
N-(4-Chlorophenyl)-1,2-phenylenediamine 99%
N-(4-Chlorophenyl)-1,2-phenylenediamine
Formula: C₁₂H₁₁ClN₂
MW: 218.682 g/mol
Melting Pt: 117…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00007686
CAS Number: 68817-71-0