4-[(1R)-1-Aminoethyl]phenol
Supplier: Apollo Scientific
Synonyms: (R)-4-Hydroxy-α-methylbenzylamine, (1R)-1-(4-Hydroxyphenyl)ethylamine
OR48265-1G OR48265-250MG
APOSOR48265-1GEA 677 EUR
APOSOR48265-1G APOSOR48265-250MG
4-[(1R)-1-Aminoethyl]phenol
4-[(1R)-1-Aminoethyl]phenol
Formula: C₈H₁₁NO
MW: 137.179 g/mol
Melting Pt: 121 °C
Storage Temperature: Ambient
MDL Number: MFCD03844647
CAS Number: 134855-88-2