(1S,3R)-Cyclopentane-1,3-diol
Supplier: Apollo Scientific
Synonyms: cis-Cyclopentane-1,3-diol
OR49056-1G OR49056-250MG
APOSOR49056-1GEA 922 EUR
APOSOR49056-1G APOSOR49056-250MG
(1S,3R)-Cyclopentane-1,3-diol
(1S,3R)-Cyclopentane-1,3-diol
Formula: C₅H₁₀O₂
MW: 102.132 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13619671
CAS Number: 16326-97-9