5-Chloropyrazolo[1,5-a]pyrimidin-3-amine
Supplier: Apollo Scientific
OR49061-1G OR49061-250MG
APOSOR49061-1GEA 106 EUR
APOSOR49061-1G APOSOR49061-250MG
5-Chloropyrazolo[1,5-a]pyrimidin-3-amine
5-Chloropyrazolo[1,5-a]pyrimidin-3-amine
Formula: C₆H₅ClN₄
MW: 168.584 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17016123
CAS Number: 1234616-50-2