6-Bromo-4-methoxy-pyrazolo[1,5-a]pyridine
Supplier: Apollo Scientific
Synonyms: 6-Bromo-4-methoxypyrazolo[1,5-a]pyridine
OR49065-1G OR49065-250MG
APOSOR49065-1GEA 235 EUR
APOSOR49065-1G APOSOR49065-250MG
6-Bromo-4-methoxy-pyrazolo[1,5-a]pyridine
6-Bromo-4-methoxy-pyrazolo[1,5-a]pyridine
Formula: C₈H₇BrN₂O
MW: 227.058 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17926335
CAS Number: 1207557-36-5