3-(Chloromethyl)benzaldehyde 97%
Supplier: Apollo Scientific
Warning
OR500005-100MG OR500005-250MG
APOSOR500005-100MGEA 241 EUR
APOSOR500005-100MG APOSOR500005-250MG
3-(Chloromethyl)benzaldehyde 97%
3-(Chloromethyl)benzaldehyde
Formula: C₈H₇ClO
MW: 154.594 g/mol
Storage Temperature: Freezer
MDL Number: MFCD19232727
CAS Number: 77072-00-5