4-Chloro-1,8-naphthyridin-2(1H)-one 95+%
Supplier: Apollo Scientific
Warning
OR500035-100MG OR500035-250MG
APOSOR500035-100MGEA 63.8 EUR
APOSOR500035-100MG APOSOR500035-250MG
4-Chloro-1,8-naphthyridin-2(1H)-one 95+%
4-Chloro-1,8-naphthyridin-2(1H)-one
Formula: C₈H₅ClN₂O
MW: 180.591 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD24556140
CAS Number: 59514-93-1