5,5-Dimethylbicyclo[4.1.0]heptan-2-one
Supplier: Apollo Scientific
OR50906-1G OR50906-250MG OR50906-5G
APOSOR50906-1GEA 511 EUR
APOSOR50906-1G APOSOR50906-250MG APOSOR50906-5G
5,5-Dimethylbicyclo[4.1.0]heptan-2-one
5,5-Dimethylbicyclo[4.1.0]heptan-2-one
Formula: C₉H₁₄O
MW: 138.207 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24691569
CAS Number: 88105-44-6
UN: 1993
ADR: 3,III